Merck
  • A self-microemulsifying drug delivery system to overcome intestinal resveratrol toxicity and presystemic metabolism.
A self-microemulsifying drug delivery system to overcome intestinal resveratrol toxicity and presystemic metabolism.
Journal of pharmaceutical sciences (2014-08-12)
Katarina Bolko Seljak, Katja Berginc, Jurij Trontelj, Alenka Zvonar, Albin Kristl, Mirjana Gašperlin
ABSTRACT
A mixed lipid-mixed surfactant self-microemulsifying drug delivery system (SMEDDS) was developed to exploit the health benefits of resveratrol, a Biopharmaceutical Classification System Class 2 natural polyphenol, subject to extensive intestinal presystemic metabolism. SMEDDS with a mixed lipid phase (castor oil/Capmul MCM 1:1) and a mixed surfactant phase (Kolliphor EL/Kolliphor RH 40 1:1) was developed and evaluated for its self-emulsifying properties and in vitro dispersion. The impact of SMEDDS on the permeability properties of resveratrol and its metabolite fluxes through the rat intestine and Caco-2 cells was monitored. The inhibitory effect of selected SMEDDS components on the efflux transporters multidrug resistance-associated protein and P-gp as well as cytotoxicity was assessed on Caco-2 cells. The formulation allowed for high resveratrol loading (122.5 mg/g SMEDDS), excellent self-emulsifying properties, and very rapid release. When formulated in SMEDDS, resveratrol metabolite efflux significantly declined. The formulation (SMEDDS without incorporated resveratrol) and its individual components did not compromise in vitro cell vitality and integrity. Mixed lipid-mixed surfactant SMEDDS is a prospective formulation to improve resveratrol biopharmaceutical, pharmacokinetic, and toxicological properties, leading the way to resveratrol use not only as a supplement but also as a pharmacological drug.
Product Description
Supelco
Water, for TOC analysis
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
1-Octanoyl-rac-glycerol, ≥99%
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, PCR Reagent
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Water, ACS reagent, for ultratrace analysis
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Water, Deionized
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Water, ACS reagent
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Resveratrol, European Pharmacopoeia (EP) Reference Standard
Supelco
Resveratrol, analytical standard
Supelco
Resveratrol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Sigma-Aldrich
Rhodamine 123, mitochondrial specific fluorescent dye
Sigma-Aldrich
Rhodamine 123, BioReagent, for fluorescence, ≥85% (HPLC)