Merck
  • Chk1 is required for the metaphase-anaphase transition via regulating the expression and localization of Cdc20 and Mad2.
Chk1 is required for the metaphase-anaphase transition via regulating the expression and localization of Cdc20 and Mad2.
Life sciences (2014-04-22)
Xiaoyun Yang, Wei Xu, Zuowei Hu, Yaou Zhang, Naihan Xu
ABSTRACT
The checkpoint kinase 1 (Chk1) functions not only in genotoxic stresses but also in normal cell cycle progression, particularly in the initiation, progression and fidelity of unperturbed mitosis. In this study, we investigated the role of Chk1 in regulating the metaphase-anaphase transition in mammalian cells. The mitotic progression was monitored by flow cytometry analysis. The levels of cyclin B1, Cdc20 and Mad2 were measured by Western blotting. Metaphase chromosome alignment and the subcellular localization of Cdc20 and Mad2 were analyzed by immunofluorescence and confocal microscopy. Cyclin B1 degradation and the metaphase-anaphase transition were severely blocked by Chk1 siRNA. Depletion of Chk1 induced chromosome alignment defect in metaphase cells. The kinetochore localization of Cdc20, Mad2 was disrupted in Chk1 depleted cells. Chk1 abrogation also dramatically reduced the protein expression levels of Cdc20 and Mad2. These results strongly suggest that Chk1 is required for the metaphase-anaphase transition via regulating the subcellular localization and the expression of Cdc20 and Mad2.
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Paclitaxel semi-synthetic for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
MISSION® esiRNA, targeting human CHKB, CHKB-CPT1B
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ckb
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder
Sigma-Aldrich
Paclitaxel, from semisynthetic, ≥98%
Sigma-Aldrich
Paclitaxel, from Taxus yannanensis, powder
Paclitaxel, European Pharmacopoeia (EP) Reference Standard
Paclitaxel semi-synthetic for system suitability, European Pharmacopoeia (EP) Reference Standard
Paclitaxel natural for peak identification, European Pharmacopoeia (EP) Reference Standard