Merck
  • Steam explosion distinctively enhances biomass enzymatic saccharification of cotton stalks by largely reducing cellulose polymerization degree in G. barbadense and G. hirsutum.
Steam explosion distinctively enhances biomass enzymatic saccharification of cotton stalks by largely reducing cellulose polymerization degree in G. barbadense and G. hirsutum.
Bioresource technology (2015-02-07)
Yu Huang, Xiaoyang Wei, Shiguang Zhou, Mingyong Liu, Yuanyuan Tu, Ao Li, Peng Chen, Yanting Wang, Xuewen Zhang, Hongzhong Tai, Liangcai Peng, Tao Xia
ABSTRACT
In this study, steam explosion pretreatment was performed in cotton stalks, leading to 5-6 folds enhancements on biomass enzymatic saccharification distinctive in Gossypium barbadense and Gossypium hirsutum species. Sequential 1% H2SO4 pretreatment could further increase biomass digestibility of the steam-exploded stalks, and also cause the highest sugar-ethanol conversion rates probably by releasing less inhibitor to yeast fermentation. By comparison, extremely high concentration alkali (16% NaOH) pretreatment with raw stalks resulted in the highest hexoses yields, but it had the lowest sugar-ethanol conversion rates. Characterization of wall polymer features indicated that biomass saccharification was enhanced with steam explosion by largely reducing cellulose DP and extracting hemicelluloses. It also showed that cellulose crystallinity and arabinose substitution degree of xylans were the major factors on biomass digestibility in cotton stalks. Hence, this study has provided the insights into cell wall modification and biomass process technology in cotton stalks and beyond.
Product Description
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Acetic anhydride, Arxada quality, ≥99.5% (GC)
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
D-(+)-Xylose, ≥99%
Sigma-Aldrich
Acetic anhydride, ACS reagent, ≥98.0%
Sigma-Aldrich
Syringaldehyde, 98%
Sigma-Aldrich
D-(+)-Xylose, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Acetic anhydride, 99.5%
Supelco
Acetic anhydride, for GC derivatization, LiChropur, ≥99.0%
Sigma-Aldrich
Acetic anhydride, ReagentPlus®, ≥99%
Sigma-Aldrich
Syringaldehyde, ≥98%, FG
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
D-(+)-Xylose, BioXtra, ≥99% (GC)
Sigma-Aldrich
D-(+)-Xylose, ≥99% (GC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
L-Rhamnose, natural sourced, 99%, FG
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard
Supelco
Vanillin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-(+)-Arabinose, 99%