Merck
  • The inhibitory effect of selenium nanoparticles on protein glycation in vitro.
The inhibitory effect of selenium nanoparticles on protein glycation in vitro.
Nanotechnology (2015-03-19)
Shaoxuan Yu, Wentao Zhang, Wei Liu, Wenxin Zhu, Ruochen Guo, Yashan Wang, Daohong Zhang, Jianlong Wang
ABSTRACT
Selenium nanoparticles (Se NPs) possess well-known excellent biological activities and low toxicity, and have been employed for numerous applications except as inhibitors to protein glycation. Herein, the present study is carried out to investigate the inhibitory effect of Se NPs on protein glycation in a bovine serum albumin (BSA)/glucose system. By measuring the amount of glucose covalently bound onto BSA, the formation of fructosamine and fluorescent products, it is found that Se NPs can hinder the development of protein glycation in a dose-dependent but time-independent manner under the selected reaction conditions (55 °C, 40 h). And after comparing the increase of inhibitory rate in different stages, it is observed that Se NPs show the greatest inhibitory effect in the early stage, then in the advanced stage, but no effect in the intermediate stage. Fourier transform infrared spectroscopy characterization of Se NPs collected after glycation and determination of ·OH influence and glyoxal formation show that the mechanism for the inhibitory efficacy of Se NPs is related to their strong competitive activity against available amino groups in proteins, their great scavenging activity on reactive oxygen species and their inhibitory effect on α-dicarbonyl compounds' formation. In addition, it is proved that Se NPs protect proteins from structural modifications in the system and they do not exhibit significant cytotoxicity towards BV-2 and BRL-3A cells at low concentrations (10 and 50 μg mL(-1)). Consequently, Se NPs may be suitable for further in vivo studies as novel anti-glycation agents.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium thiosulfate pentahydrate, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
N,N′-Methylenebis(acrylamide), 99%
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
Phthaldialdehyde, for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
N,N′-Methylenebisacrylamide, suitable for electrophoresis (after filtration or allowing insolubles to settle)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Sigma-Aldrich
Phthaldialdehyde, ≥97% (HPLC), powder or crystals
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Sigma-Aldrich
Sodium thiosulfate pentahydrate, BioXtra, ≥99.5%
Sigma-Aldrich
Phthaldialdehyde, suitable for HPLC fluorimetric detection of amino acids, ≥99% (HPLC), powder or crystals
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
N,N′-Methylenebisacrylamide, powder, for molecular biology, suitable for electrophoresis, ≥99.5%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)