Merck
  • ENMD-1068, a protease-activated receptor 2 antagonist, inhibits the development of endometriosis in a mouse model.
ENMD-1068, a protease-activated receptor 2 antagonist, inhibits the development of endometriosis in a mouse model.
American journal of obstetrics and gynecology (2014-02-06)
Yifeng Wang, Min Lin, Huinan Weng, Xuefeng Wang, Li Yang, Fenghua Liu
ABSTRACT
Protease-activated receptor 2 plays an important role in the pathogenesis of endometriosis. We studied the effect of ENMD-1068, a protease-activated receptor 2 antagonist, on the development of endometriosis in a noninvasive fluorescent mouse model. A red fluorescent protein-expressing xenograft model of human endometriosis was created in nude mice. After endometriosis induction, the mice were injected intraperitoneally with either 25 mg/kg or 50 mg/kg ENMD-1068 or with 200 μL of the vehicle control daily for 5 days. The endometriotic lesions that developed in the mice were then counted, measured, and collected. The lesions were assessed for the production of interleukin 6 and monocyte chemotactic protein-1 by enzyme-linked immunosorbent assays and evaluated for the activation of nuclear factor-κB and the expression of vascular endothelial growth factor by immunohistochemical analyses. Cell proliferation and apoptosis were assessed by immunohistochemistry for Ki-67 and terminal deoxynucleotidyl transferase-mediated dUTP nick end labeling, respectively. ENMD-1068 dose-dependently inhibited the development of endometriotic lesions (P < .05) without apparent toxicity to various organs of the treated mice. Consistently, ENMD-1068 dose-dependently inhibited the expression of interleukin 6 and nuclear factor-κB (P < .05) and cell proliferation (P < .05) in the lesions, as well as increased the percentage of apoptotic cells (P < .05). ENMD-1068 reduced the levels of monocyte chemotactic protein-1 and vascular endothelial growth factor in the lesions (P < .05), but not in a dose-dependent manner. Our study suggests that ENMD-1068 is effective in suppressing the growth of endometriosis, which might be attributed to the drug's antiangiogenic and antiinflammatory activities.
Product Description
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Sigma-Aldrich
ENMD-1068 hydrochloride, ≥97% (HPLC)