Merck
  • Overexpression of MEOX2 and TWIST1 is associated with H3K27me3 levels and determines lung cancer chemoresistance and prognosis.
Overexpression of MEOX2 and TWIST1 is associated with H3K27me3 levels and determines lung cancer chemoresistance and prognosis.
PloS one (2014-12-03)
Federico Ávila-Moreno, Leonel Armas-López, Aldo M Álvarez-Moran, Zoila López-Bujanda, Blanca Ortiz-Quintero, Alfredo Hidalgo-Miranda, Francisco Urrea-Ramírez, R María Rivera-Rosales, Eugenia Vázquez-Manríquez, Erika Peña-Mirabal, José Morales-Gómez, Juan C Vázquez-Minero, José L Téllez-Becerra, Roberto Ramírez-Mendoza, Alejandro Ávalos-Bracho, Enrique Guzmán de Alba, Karla Vázquez-Santillán, Vilma Maldonado-Lagunas, Patricio Santillán-Doherty, Patricia Piña-Sánchez, Joaquin Zúñiga-Ramos
ABSTRACT
Lung cancer is the leading cause of death from malignant diseases worldwide, with the non-small cell (NSCLC) subtype accounting for the majority of cases. NSCLC is characterized by frequent genomic imbalances and copy number variations (CNVs), but the epigenetic aberrations that are associated with clinical prognosis and therapeutic failure remain not completely identify. In the present study, a total of 55 lung cancer patients were included and we conducted genomic and genetic expression analyses, immunohistochemical protein detection, DNA methylation and chromatin immunoprecipitation assays to obtain genetic and epigenetic profiles associated to prognosis and chemoresponse of NSCLC patients. Finally, siRNA transfection-mediated genetic silencing and cisplatinum cellular cytotoxicity assays in NSCLC cell lines A-427 and INER-37 were assessed to describe chemoresistance mechanisms involved. Our results identified high frequencies of CNVs (66-51% of cases) in the 7p22.3-p21.1 and 7p15.3-p15.2 cytogenetic regions. However, overexpression of genes, such as MEOX2, HDAC9, TWIST1 and AhR, at 7p21.2-p21.1 locus occurred despite the absence of CNVs and little changes in DNA methylation. In contrast, the promoter sequences of MEOX2 and TWIST1 displayed significantly lower/decrease in the repressive histone mark H3K27me3 and increased in the active histone mark H3K4me3 levels. Finally these results correlate with poor survival in NSCLC patients and cellular chemoresistance to oncologic drugs in NSCLC cell lines in a MEOX2 and TWIST1 overexpression dependent-manner. In conclusion, we report for the first time that MEOX2 participates in chemoresistance irrespective of high CNV, but it is significantly dependent upon H3K27me3 enrichment probably associated with aggressiveness and chemotherapy failure in NSCLC patients, however additional clinical studies must be performed to confirm our findings as new probable clinical markers in NSCLC patients.
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Supelco
Acrylamide, analytical standard
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Acrylamide, for molecular biology, ≥99% (HPLC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%