Merck
  • RAGE influences the development of aortic valve stenosis in mice on a high fat diet.
RAGE influences the development of aortic valve stenosis in mice on a high fat diet.
Experimental gerontology (2014-05-14)
Britt Hofmann, Yuliana Yakobus, Mailiza Indrasari, Norbert Nass, Alexander Navarrete Santos, Frank Bernhard Kraus, Rolf-Edgar Silber, Andreas Simm
ABSTRACT
Advanced glycation end product (AGE) accumulations as well as a high fat diet are associated with cardiovascular diseases. AGEs are recognized by several receptor molecules of which the receptor of AGEs (RAGE) is currently the most intensively studied. Activation of RAGE causes an unfavorable pro-inflammatory state. The hypothesis of this study was that metabolic stress due to a high fat diet results in the development of aortic valve stenosis and that knockout of RAGE should be protective. Six week old male C57BL/6N and C57BL/6N RAGE-/- mice (n=28) were randomly assigned to 4 groups and fed with normal or high fat diet for 32weeks. Weight gain was determined weekly. At the start of the experiment and after 2, 4 and 7months, echocardiographic assessments of the aortic valve were made. At the end of the experiment, plasma lipid levels and histological changes of the valves were determined. The high fat diet resulted in accelerated weight gain. However, after 7month, only C57BL/6 mice developed increased trans-aortic-valve velocities, leaflet thickness and reduced valve area index (p<0.0001). Immunohistochemistry of the aortic valves revealed in C57BL/6N mice on a high fat diet more calcification, AGE accumulation and RAGE expression when compared to normal fed control. Hearts and aortic valves of RAGE-/- mice showed less morphometric changes, calcification and AGE accumulation. After 7months of high fat feeding C57BL/6 mice (p<0.0001) as well as RAGE-/- mice (p=0.007) had significantly increased cholesterol levels compared to normal fed control, however RAGE-/- mice were probably protected due to a better HDL/LDL ratio when compared to wild type animals (p=0.003). These data suggest that AGEs and RAGE are involved in the development of obesity, hypercholesterolemia and aortic valve changes due to metabolic stress from high fat intake.
Product Description
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
SAFC
Cholesterol, Plant-Derived, SyntheChol®