Merck
  • Colocalization, physical, and functional interaction between Werner and Bloom syndrome proteins.
Colocalization, physical, and functional interaction between Werner and Bloom syndrome proteins.
The Journal of biological chemistry (2002-03-29)
Cayetano von Kobbe, Parimal Karmakar, Lale Dawut, Patricia Opresko, Xianmin Zeng, Robert M Brosh, Ian D Hickson, Vilhelm A Bohr
ABSTRACT
The RecQ helicase family comprises a conserved group of proteins implicated in several aspects of DNA metabolism. Three of the family members are defective in heritable diseases characterized by abnormal growth, premature aging, and predisposition to malignancies. These include the WRN and BLM gene products that are defective in Werner and Bloom syndromes, disorders which share many phenotypic and cellular characteristics including spontaneous genomic instability. Here, we report a physical and functional interaction between BLM and WRN. These proteins were coimmunoprecipitated from a nuclear matrix-solubilized fraction, and the purified recombinant proteins were shown to interact directly. Moreover, BLM and WRN colocalized to nuclear foci in three human cell lines. Two regions of WRN that mediate interaction with BLM were identified, and one of these was localized to the exonuclease domain of WRN. Functionally, BLM inhibited the exonuclease activity of WRN. This is the first demonstration of a physical and functional interaction between RecQ helicases. Our observation that RecQ family members interact provides new insights into the complex phenotypic manifestations resulting from the loss of these proteins.