Merck
Porphyrin-based switchable molecular turnstiles.
Chemistry (Weinheim an der Bergstrasse, Germany) (2011-04-22)
Aurélie Guenet, Ernest Graf, Nathalie Kyritsakas, Mir Wais Hosseini
ABSTRACT
The design, synthesis and structural characterisation, in solution, of two new molecular turnstiles based on Sn-porphyrin derivatives are described. The system is composed of a stator (5-(4-pyridyl)-10,15,20-triphenylporphyrin), a hinge (Sn(IV)) and a rotor (handle equipped with 2,6-pyridinedicarboxamide as a tridentate coordinating site or its Pd(II) complex). The presence of interaction sites, both on the stator and the rotor, offers the possibility of switching between an open state (free rotation of the handle around the porphyrin) and a closed state (blockage of the rotation) by either establishment of hydrogen bonds between the stator and the rotor or by the simultaneous binding of Pd by both coordinating groups.
Sigma-Aldrich
2,6-Pyridinedicarboxamide, 97%