Merck
  • A rice-derived recombinant human lactoferrin stimulates fibroblast proliferation, migration, and sustains cell survival.
A rice-derived recombinant human lactoferrin stimulates fibroblast proliferation, migration, and sustains cell survival.
Wound repair and regeneration : official publication of the Wound Healing Society [and] the European Tissue Repair Society (2010-01-20)
Ling Tang, Tengjiao Cui, James J Wu, Wen Liu-Mares, Ning Huang, Jie Li
ABSTRACT
Human lactoferrin (hLF), a glycoprotein of the transferrin family, has recently been shown to stimulate wound repair through its antimicrobial effect and inflammation modulation. A recent study with several non-skin cell lines indicated that hLF may also have a stimulatory effect on cell proliferation. To explore the role of hLF in wound healing, we used recombinant human lactoferrin (holo-rhLF), derived from transgenic rice, to examine the effects of holo-rhLF on cell proliferation, migration, attachment, and survival in a human primary skin fibroblast culture system. This study revealed that holo-rhLF not only significantly stimulates fibroblast proliferation but also has synergistic effects with fibroblast growth factor-2 and antagonistic effects with transforming growth factor-beta1 on cell proliferation. Furthermore, using a chamber migration assay, our results demonstrate that holo-rhLF promotes fibroblast migration in a dosage-dependent manner. More importantly, holo-rhLF significantly increased cell viability and protected cells from death when they were stressed by either serum depletion or 12-O-tetradecanoylphorbol-13-acetate exposure. No significant effect was observed on cell attachment. In conclusion, these findings reveal the multiple functions of holo-rhLF in human skin fibroblasts and indicate its potential application in wound therapy by enhancing cell proliferation and migration as well as protecting cells from apoptosis.
Sigma-Aldrich
Lactoferrin human, recombinant, expressed in rice, Iron saturated, ≥90% (SDS-PAGE)
Pricing and availability is not currently available.