Merck
  • Optimization of a preparative capillary gas chromatography-mass spectrometry system for the isolation and harvesting of individual polycyclic aromatic hydrocarbons.
Optimization of a preparative capillary gas chromatography-mass spectrometry system for the isolation and harvesting of individual polycyclic aromatic hydrocarbons.
Journal of chromatography. A (2003-07-02)
Manolis Mandalakis, Orjan Gustafsson
ABSTRACT
Operation parameters of a preparative capillary gas chromatography (pcGC) system were optimized to facilitate clean and efficient harvesting of individual polycyclic aromatic hydrocarbons (PAHs) for subsequent compound-specific radiocarbon analysis. For PAHs, the recommended optimized settings of the specially-designed pcGC cooled injection system (CIS) and preparative fraction collector (PFC) are: 5 s CIS solvent venting time, deactivation of CIS "stop flow" injection mode, autoinjector "fast injection" mode, 60 s CIS splitless time, 340 degrees C PFC switch temperature, and 30 degrees C (ambient) trapping temperature. These optimized conditions yielded highly reproducible, pure, and efficient pcGC harvesting of six PAHs with mass recoveries of 90-100% and purity of the isolates of 97-100%.
Product Description
Supelco
SPB®-5 Capillary GC Column, L × I.D. 50 m × 0.32 mm, df 5.00 μm
Pricing and availability is not currently available.