Merck
  • Domain and functional analysis of a novel platelet-endothelial cell surface protein, SCUBE1.
Domain and functional analysis of a novel platelet-endothelial cell surface protein, SCUBE1.
The Journal of biological chemistry (2008-02-28)
Cheng-Fen Tu, Yu-Ting Yan, Szu-Yao Wu, Bambang Djoko, Ming-Tzu Tsai, Chien-Jui Cheng, Ruey-Bing Yang
ABSTRACT
SCUBE1 (signal peptide-CUB-EGF domain-containing protein 1) is a novel, secreted, cell surface glycoprotein expressed during early embryogenesis and found in platelet and endothelial cells. This protein is composed of an N-terminal signal peptide sequence followed by nine tandemly arranged epidermal growth factor (EGF)-like repeats, a spacer region, three cysteine-rich repeat motifs, and one CUB domain at the C terminus. However, little is known about its domain and biological function. Here, we generated a comprehensive panel of domain deletion constructs and a new genetic mouse model with targeted disruption of Scube1 (Scube1(Delta cub/Delta cub)) to investigate the domain function and biological significance. A number of cell-based assays were utilized to define the critical role of the spacer region for membrane association and establish that the EGF-like repeats 7-9 are sufficient for the formation of SCUBE1-mediated homophilic adhesions in a calcium-dependent fashion. Biochemical and molecular analyses showed that the C-terminal cysteine-rich motifs and CUB domain could directly bind and antagonize the bone morphogenetic protein activity. Furthermore, genetic ablation of this C-terminal region resulted in brain malformation in the Scube1(Delta cub/Delta cub) embryos. Together, our results support the dual roles of SCUBE1 on brain morphogenesis and cell-cell adhesions through its distinct domain function.