Merck
  • Leptin enhances insulin sensitivity by direct and sympathetic nervous system regulation of muscle IGFBP-2 expression: evidence from nonrodent models.
Leptin enhances insulin sensitivity by direct and sympathetic nervous system regulation of muscle IGFBP-2 expression: evidence from nonrodent models.
Endocrinology (2014-03-25)
Steven W Yau, Belinda A Henry, Vincenzo C Russo, Glenn K McConell, Iain J Clarke, George A Werther, Matthew A Sabin
ABSTRACT
Leptin is produced from white adipose tissue and acts primarily to regulate energy balance. Obesity is associated with leptin resistance and increased circulating levels of leptin. Leptin has recently been shown to influence levels of IGF binding protein-2 (IGFBP-2), a protein that is reduced in obesity and type 2 diabetes. Overexpression of IGFBP-2 protects against obesity and type 2 diabetes. As such, IGFBP-2 signaling may represent a novel pathway by which leptin regulates insulin sensitivity. We sought to investigate how leptin regulates skeletal muscle IGFBP-2 levels and to assess the impact of this on insulin signaling and glucose uptake. In vitro experiments were undertaken in cultured human skeletal myotubes, whereas in vivo experiments assessed the effect of intracerebroventricular leptin on peripheral skeletal muscle IGFBP-2 expression and insulin sensitivity in sheep. Leptin directly increased IGFBP-2 mRNA and protein in human skeletal muscle through both signal transducer and activator of transcription-3 and phosphatidylinositol 3-kinase signaling, in parallel with enhanced insulin signaling. Silencing IGFBP-2 lowered leptin- and insulin-stimulated protein kinase B phosphorylation and glucose uptake. In in vivo experiments, intracerebroventricular leptin significantly increased hind-limb skeletal muscle IGFBP-2, an effect completely blocked by concurrent peripheral infusion of a β-adrenergic blocking agent. Sheep receiving central leptin showed improvements in glucose tolerance and circulating insulin levels after an iv glucose load. In summary, leptin regulates skeletal muscle IGFBP-2 by both direct peripheral and central (via the sympathetic nervous system) mechanisms, and these likely impact on peripheral insulin sensitivity and glucose metabolism.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Pricing and availability is not currently available.
SAFC
Sodium chloride solution, 5 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Leptin human, ≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 5 M
Pricing and availability is not currently available.
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, tablet
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Pricing and availability is not currently available.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Pricing and availability is not currently available.
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human IGFBP2
Pricing and availability is not currently available.
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Pricing and availability is not currently available.
Supelco
Dimethyl sulfoxide, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Pricing and availability is not currently available.