Merck
  • Acetyl-CoA synthetase 2 promotes acetate utilization and maintains cancer cell growth under metabolic stress.
Acetyl-CoA synthetase 2 promotes acetate utilization and maintains cancer cell growth under metabolic stress.
Cancer cell (2015-01-15)
Zachary T Schug, Barrie Peck, Dylan T Jones, Qifeng Zhang, Shaun Grosskurth, Israt S Alam, Louise M Goodwin, Elizabeth Smethurst, Susan Mason, Karen Blyth, Lynn McGarry, Daniel James, Emma Shanks, Gabriela Kalna, Rebecca E Saunders, Ming Jiang, Michael Howell, Francois Lassailly, May Zaw Thin, Bradley Spencer-Dene, Gordon Stamp, Niels J F van den Broek, Gillian Mackay, Vinay Bulusu, Jurre J Kamphorst, Saverio Tardito, David Strachan, Adrian L Harris, Eric O Aboagye, Susan E Critchlow, Michael J O Wakelam, Almut Schulze, Eyal Gottlieb
ABSTRACT
A functional genomics study revealed that the activity of acetyl-CoA synthetase 2 (ACSS2) contributes to cancer cell growth under low-oxygen and lipid-depleted conditions. Comparative metabolomics and lipidomics demonstrated that acetate is used as a nutritional source by cancer cells in an ACSS2-dependent manner, and supplied a significant fraction of the carbon within the fatty acid and phospholipid pools. ACSS2 expression is upregulated under metabolically stressed conditions and ACSS2 silencing reduced the growth of tumor xenografts. ACSS2 exhibits copy-number gain in human breast tumors, and ACSS2 expression correlates with disease progression. These results signify a critical role for acetate consumption in the production of lipid biomass within the harsh tumor microenvironment.
Sigma-Aldrich
S-Acetyl-coenzyme A synthetase from baker′s yeast (S. cerevisiae), lyophilized powder, ≥3 units/mg protein
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-ACSS2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-ACSS1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.