Merck
  • Personalised chemotherapy based on tumour marker decline in poor prognosis germ-cell tumours (GETUG 13): a phase 3, multicentre, randomised trial.
Personalised chemotherapy based on tumour marker decline in poor prognosis germ-cell tumours (GETUG 13): a phase 3, multicentre, randomised trial.
The Lancet. Oncology (2014-12-03)
Karim Fizazi, Lance Pagliaro, Agnes Laplanche, Aude Fléchon, Josef Mardiak, Lionnel Geoffrois, Pierre Kerbrat, Christine Chevreau, Remy Delva, Frederic Rolland, Christine Theodore, Guilhem Roubaud, Gwenaëlle Gravis, Jean-Christophe Eymard, Jean-Pierre Malhaire, Claude Linassier, Muriel Habibian, Anne-Laure Martin, Florence Journeau, Maria Reckova, Christopher Logothetis, Stephane Culine
ABSTRACT
Poor prognosis germ-cell tumours are only cured in about half of patients. We aimed to assess whether treatment intensification based on an early tumour marker decline will improve progression-free survival for patients with germ-cell tumours. In this phase 3, multicentre, randomised trial, patients were enrolled from France (20 centres), USA (one centre), and Slovakia (one centre). Patients were eligible if they were older than 16 years, had evidence of testicular, retroperitoneal, or mediastinal non-seminomatous germ cell tumours based on histological findings or clinical evidence and highly elevated serum human chorionic gonadotropin or alfa-fetoprotein concentrations that matched International Germ Cell Cancer Consensus Group poor prognosis criteria. After one cycle of BEP (intravenous cisplatin [20 mg/m(2) per day for 5 days], etoposide [100 mg/m(2) per day for 5 days], and intramuscular or intravenous bleomycin [30 mg per day on days 1, 8, and 15]), patients' human chorionic gonadotropin and alfa-fetoprotein concentrations were measured at day 18-21. Patients with a favourable decline in human chorionic gonadotropin and alfa-fetoprotein continued BEP (Fav-BEP group) for 3 additonal cycles, whereas patients with an unfavourable decline were randomly assigned (1:1) to receive either BEP (Unfav-BEP group) or a dose-dense regimen (Unfav-dose-dense group), consisting of intravenous paclitaxel (175 mg/m(2) over 3 h on day 1) before BEP plus intravenous oxaliplatin (130 mg/m(2) over 3 h on day 10; two cycles), followed by intravenous cisplatin (100 mg/m(2) over 2 h on day 1), intravenous ifosfamide (2 g/m(2) over 3 h on days 10, 12, and 14), plus mesna (500 mg/m(2) at 0, 3, 7 and 11 h), and bleomycin (25 units per day, by continuous infusion for 5 days on days 10-14; two cycles), with granulocyte-colony stimulating factor (lenograstim) support. Centrally blocked computer-generated randomisation stratified by centre was used. The primary endpoint was progression-free survival and the efficacy analysis was done in the intention-to-treat population. The planned trial accrual was completed in May, 2012, and follow-up is ongoing. This study is registered with ClinicalTrials.gov, number NCT00104676. Between Nov 28, 2003, and May 16, 2012, 263 patients were enrolled and 254 were available for tumour marker assessment. Of these 51 (20%) had a favourable marker assessment, and 203 (80%) had an unfavourable tumour marker decline; 105 were randomly assigned to the Unfav-dose-dense group and 98 to the Unfav-BEP group. 3-year progression-free survival was 59% (95% CI 49-68) in the Unfav-dose-dense group versus 48% (38-59) in the Unfav-BEP group (HR 0·66, 95% CI 0·44-1·00, p=0·05). 3-year progression-free survival was 70% (95% CI 57-81) in the Fav-BEP group (HR 0·66, 95% CI 0·49-0·88, p=0·01 for progression-free survival compared with the Unfav-BEP group). More grade 3-4 neurotoxic events (seven [7%] vs one [1%]) and haematotoxic events occurred in the Unfav-dose-dense group compared with in the Unfav-BEP group; there was no difference in grade 1-2 febrile neutropenia (18 [17%] vs 18 [18%]) or toxic deaths (one [1%] in both groups). Salvage high-dose chemotherapy plus a stem-cell transplant was required in six (6%) patients in the Unfav-dose-dense group and 16 (16%) in the Unfav-BEP group. Personalised treatment with chemotherapy intensification reduces the risk of progression or death in patients with poor prognosis germ-cell tumours and an unfavourable tumour marker decline. Institut National du Cancer (Programme Hospitalier de Recherche Clinique).
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Paclitaxel semi-synthetic for peak identification, European Pharmacopoeia (EP) Reference Standard
Bleomycin sulfate, European Pharmacopoeia (EP) Reference Standard
Etoposide for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ifosfamide, ≥98%
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, BioXtra, crystalline
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, for fluorescence, mixture of bleomycin sulfate salts, lyophilized, powder or crystals, white to off-white
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, 1.5-2.0 units/mg solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Paclitaxel, from semisynthetic, ≥98%
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, crystalline, 1.5-2.0 U/mg
Sigma-Aldrich
Paclitaxel, from Taxus yannanensis, powder
Sigma-Aldrich
Oxaliplatin, powder
Paclitaxel semi-synthetic for system suitability, European Pharmacopoeia (EP) Reference Standard
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Paclitaxel, European Pharmacopoeia (EP) Reference Standard
Ifosfamide, European Pharmacopoeia (EP) Reference Standard
Oxaliplatin, European Pharmacopoeia (EP) Reference Standard
Etoposide, European Pharmacopoeia (EP) Reference Standard
Paclitaxel natural for peak identification, European Pharmacopoeia (EP) Reference Standard