Merck
  • Association of matrilysin-2 (MMP-26) expression with tumor progression and activation of MMP-9 in esophageal squamous cell carcinoma.
Association of matrilysin-2 (MMP-26) expression with tumor progression and activation of MMP-9 in esophageal squamous cell carcinoma.
Carcinogenesis (2004-08-31)
Hiroyuki Yamamoto, Akravit Vinitketkumnuen, Yasushi Adachi, Hiroaki Taniguchi, Tamaki Hirata, Nobuki Miyamoto, Katsuhiko Nosho, Arisa Imsumran, Masahiro Fujita, Masao Hosokawa, Yuji Hinoda, Kohzoh Imai
ABSTRACT
Expression of matrilysin-2, matrix metalloproteinase (MMP)-26, has been implicated in the progression of several types of human cancer. Matrilysin-2 has been reported to be a physiological and pathological activator of pro-MMP-9. The aim of this study was to examine matrilysin-2 expression and determine whether it is correlated with progression of human esophageal squamous cell carcinoma (ESCC). Semi-quantitative reverse transcriptase-polymerase chain reaction, immunohistochemical analysis, zymography and an in vitro invasion assay were performed. Matrilysin-2 mRNA expression was undetectable or only faintly detected in non-tumor tissues, but its overexpression was detected in 24 of the 50 ESCC tissues. Matrilysin-2 overexpression was significantly correlated with depth of invasion, lymph node metastasis and an advance in pathological tumor node metastasis (pTNM) stage. Sections with immunostaining signals in >10% of carcinoma cells at the invasive front, which were observed in 46 of 100 cases, were judged to be positive for matrilysin-2 expression. Matrilysin-2 expression was significantly correlated with depth of invasion, lymph node and distant metastasis, advance in pTNM stage and recurrence. Expression of matrilysin-2 was significantly correlated with nuclear beta-catenin expression and MMP-9 expression. Patients with matrilysin-2-positive cancer had significantly shorter overall and disease-free survival periods than did those with matrilysin-2-negative cancer. Matrilysin-2 expression retained its significant predictive value for overall and disease-free survival in multivariate analysis. Moreover, patients with concomitant expression of matrilysin-2 and MMP-9 had the worst prognosis. Zymography revealed that matrilysin-2 expression was significantly correlated with expression of active MMP-9 in ESCC tissues. Matrilysin-2-transfected TE-1 ESCC cells showed active MMP-9 activity and were more invasive in vitro compared with mock-transfected TE-1 cells. The results of this study suggest that matrilysin-2, the expression of which is closely correlated with nuclear beta-catenin expression and active MMP-9 activity, plays a key role in the progression of ESCC.