Merck
  • Canalicular immunostaining of aminopeptidase N (CD13) as a diagnostic marker for hepatocellular carcinoma.
Canalicular immunostaining of aminopeptidase N (CD13) as a diagnostic marker for hepatocellular carcinoma.
Journal of clinical pathology (2005-09-29)
C Röcken, J Licht, A Roessner, S Carl-McGrath
ABSTRACT
Aminopeptidase N (CD13) is expressed in normal and neoplastic liver tissue, where it exhibits a characteristic canalicular pattern (CD13(can)), similar to that seen for CD10 and when antibodies crossreact with biliary glycoprotein I (p-CEA). To compare the putative diagnostic use of CD13(can) in differentiating between hepatocellular (HCC) and non-hepatocellular carcinomas metastatic to the liver (non-HCC). A retrospective study comparing 53 HCC specimens with 32 non-HCC specimens. Immunostaining was performed with HepPar1 and antibodies directed against CD10, CD13, p-CEA, and alpha fetoprotein (AFP). In the HCC group, a canalicular staining pattern was found for CD13, p-CEA, and CD10 in 51, 43, and 33 specimens, respectively. HepPar1 was positive in 29 and AFP in 17 HCC specimens. In the non-HCC group, canalicular immunostaining for CD10 and p-CEA was confined to non-neoplastic liver tissue. One poorly differentiated cholangiocarcinoma showed apical expression of CD13, resembling to some extent CD13(can). Sensitivity and specificity were 96.2% and 97.0%, respectively, for CD13(can), 81.1% and 100% for p-CEA(can), 62.3% and 100%, for CD10(can), 54.7% and 99.9% for HepPar1, and 32.1% and 100% for AFP. These results show that CD13(can) is more sensitive in differentiating between HCC and non-HCC than CD10(can), p-CEA(can), HepPar1, and AFP.