- A
ABSTRACT
The effect of addition of sodium anthranilate to 5 mM micellar solutions of gemini surfactant 1,4-bis(
Product Description
Pricing and availability is not currently available.
The effect of addition of sodium anthranilate to 5 mM micellar solutions of gemini surfactant 1,4-bis(