Merck
The human UTY gene encodes a novel HLA-B8-restricted H-Y antigen.
Journal of immunology (Baltimore, Md. : 1950) (2000-02-29)
E H Warren, M A Gavin, E Simpson, P Chandler, D C Page, C Disteche, K A Stankey, P D Greenberg, S R Riddell
ABSTRACT
The mammalian Y chromosome encodes male-specific minor histocompatibility (H-Y) Ags that are recognized by female T cells in an MHC-restricted manner. Two human H-Y epitopes presented by HLA-A2 and HLA-B7, respectively, have been identified previously and both are derived from the SMCY gene. We previously isolated CD8+ CTL clones that recognized a male-specific minor histocompatibility Ag presented by HLA-B8. In contrast to the SMCY-encoded H-Y epitopes, the B8/H-Y Ag was not presented by fibroblasts from male donors, suggesting that it was encoded by a novel gene. We now report that the HLA-B8-restricted H-Y epitope is defined by the octameric peptide LPHNHTDL corresponding to aa residues 566-573 of the human UTY protein. Transcription of the UTY gene is detected in a wide range of human tissues, but presentation of the UTY-derived H-Y epitope to CTL by cultured human cells shows significant cell-type specificity. Identification of this CTL-defined H-Y epitope should facilitate analysis of its contribution to graft/host interactions following sex-mismatched organ and bone marrow transplantation.