Merck
Organocatalyzed stereospecific C-C bond formation of β-lactams.
Organic & biomolecular chemistry (2013-11-13)
Sachin A Pawar, Saba Alapour, Sibusiso Khanyase, Zamani E D Cele, Srinivas Chitti, Hendrik G Kruger, Thavendran Govender, Per I Arvidsson
ABSTRACT
Herein, we report the development of mild, organocatalyzed routes to novel carbapenam derivatives through aldol, Mannich and Michael C-C bond forming reactions.
Sigma-Aldrich
trans-4-Methoxy-β-nitrostyrene, 99%
Pricing and availability is not currently available.