Merck
  • Deleterious effects of benomyl and carbendazim on human placental trophoblast cells.
Deleterious effects of benomyl and carbendazim on human placental trophoblast cells.
Reproductive toxicology (Elmsford, N.Y.) (2014-12-23)
Jinghua Zhou, Kang Xiong, Ye Yang, Xiaoqing Ye, Jing Liu, Feixue Li
ABSTRACT
Benomyl and carbendazim are benzimidazole fungicides that are used throughout the world against a wide range of fungal diseases of agricultural products. There is as yet little information regarding the toxicity of benzimidazole fungicides to human placenta. In this study, we utilized human placental trophoblast cell line HTR-8/SVneo (HTR-8) to access the toxic effects of benomyl and carbendazim. Our data showed that these two fungicides decreased cell viability and the percentages of cells in G0/G1 phase, as well as induced apoptosis of HTR-8 cells. The invasion and migration of HTR-8 cells were significantly inhibited by benomyl and carbendazim. We further found that benomyl and carbendazim altered the expression of protease systems (MMPs/TIPMs and uPA/PAI-1) and adhesion molecules (integrin α5 and β1) in HTR-8 cells. Our present study firstly shows the deleterious effects of benomyl and carbendazim on placental cells and suggests a potential risk of benzimidazole fungicides to human reproduction.
Sigma-Aldrich
Carbendazim, 97%