Merck
  • Non-targeted metabolomic reveals the effect of salt stress on global metabolite of halotolerant yeast Candida versatilis and principal component analysis.
Non-targeted metabolomic reveals the effect of salt stress on global metabolite of halotolerant yeast Candida versatilis and principal component analysis.
Journal of industrial microbiology & biotechnology (2014-08-03)
Wei Qi, Zhen-Chuan Fan, Chun-Ling Wang, Li-Hua Hou, Jin-Fu Liu, Xiao-Hong Cao
ABSTRACT
As one of the major microbes in the soy sauce fermentation, Candida versatilis enriches the flavor and improves the quality of soy sauce. In this study, a combination of five different GC-MS and LC-MS-based metabolome analytical approaches was used to analyze the intracellular, extracellular and whole metabolites of C. versatilis. Our results found out that a total of 132, 244 and 267 different metabolites were detectable from the intracellular, extracellular and whole part, respectively. When exposed to 0. 9 and 18 % salt, respectively, 114, 123 and 129 different intracellular metabolites, 184, 200 and 178 extracellular metabolites and 177, 188 and 186 whole metabolites were detected, respectively. Our data showed that salt enhances the metabolic capacity of C. versatilis, especially its amino acid and enhances the synthesis and secretion of some metabolites of C. versatilis, especially the aldehydes and phenols, such as vanillin, guaiacol and 5-hydroxymethylfurfural. Our data also showed that special attention has to be paid to the generation of biogenic amines when C. versatilis was treated with salt.
Sigma-Aldrich
Furfural, 99%
Sigma-Aldrich
Furfural, natural, ≥98%, FCC, FG
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Guaiacol, natural, ≥99%, FG
Sigma-Aldrich
5-Hydroxymethyl-2-furaldehyde, 99%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
5-(Hydroxymethyl)furfural, ≥99%, FG
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Furfural, ACS reagent, 99%
Sigma-Aldrich
Furfural, ≥98%, FCC, FG
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Guaiacol, oxidation indicator
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Guaiacol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Furfural, analytical standard
Supelco
5-(Hydroxymethyl)furfural, analytical standard
SAFC
Sodium chloride solution, 5 M
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material