Merck
  • Probing the functional equivalence of otoferlin and synaptotagmin 1 in exocytosis.
Probing the functional equivalence of otoferlin and synaptotagmin 1 in exocytosis.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2011-04-01)
Ellen Reisinger, Chris Bresee, Jakob Neef, Ramya Nair, Kirsten Reuter, Anna Bulankina, Régis Nouvian, Manuel Koch, Johanna Bückers, Lars Kastrup, Isabelle Roux, Christine Petit, Stefan W Hell, Nils Brose, Jeong-Seop Rhee, Sebastian Kügler, John V Brigande, Tobias Moser
ABSTRACT
Cochlear inner hair cells (IHCs) use Ca(2+)-dependent exocytosis of glutamate to signal sound information. Otoferlin (Otof), a C(2) domain protein essential for IHC exocytosis and hearing, may serve as a Ca(2+) sensor in vesicle fusion in IHCs that seem to lack the classical neuronal Ca(2+) sensors synaptotagmin 1 (Syt1) and Syt2. Support for the Ca(2+) sensor of fusion hypothesis for otoferlin function comes from biochemical experiments, but additional roles in late exocytosis upstream of fusion have been indicated by physiological studies. Here, we tested the functional equivalence of otoferlin and Syt1 in three neurosecretory model systems: auditory IHCs, adrenal chromaffin cells, and hippocampal neurons. Long-term and short-term ectopic expression of Syt1 in IHCs of Otof (-/-) mice by viral gene transfer in the embryonic inner ear and organotypic culture failed to rescue their Ca(2+) influx-triggered exocytosis. Conversely, virally mediated overexpression of otoferlin did not restore phasic exocytosis in Syt1-deficient chromaffin cells or neurons but enhanced asynchronous release in the latter. We further tested exocytosis in Otof (-/-) hippocampal neurons and in Syt1(-/-) IHCs but found no deficits in vesicle fusion. Expression analysis of different synaptotagmin isoforms indicated that Syt1 and Syt2 are absent from mature IHCs. Our data argue against a simple functional equivalence of the two C(2) domain proteins in exocytosis of IHC ribbon synapses, chromaffin cells, and hippocampal synapses.
Sigma-Aldrich
Abberior® STAR RED NHS, ≥80.0% (degree of coupling)