Merck
  • Structural similarity between defense peptide from wheat and scorpion neurotoxin permits rational functional design.
Structural similarity between defense peptide from wheat and scorpion neurotoxin permits rational functional design.
The Journal of biological chemistry (2014-03-29)
Antonina A Berkut, Dinara R Usmanova, Steve Peigneur, Peter B Oparin, Konstantin S Mineev, Tatyana I Odintsova, Jan Tytgat, Alexander S Arseniev, Eugene V Grishin, Alexander A Vassilevski
ABSTRACT
In this study, we present the spatial structure of the wheat antimicrobial peptide (AMP) Tk-AMP-X2 studied using NMR spectroscopy. This peptide was found to adopt a disulfide-stabilized α-helical hairpin fold and therefore belongs to the α-hairpinin family of plant defense peptides. Based on Tk-AMP-X2 structural similarity to cone snail and scorpion potassium channel blockers, a mutant molecule, Tk-hefu, was engineered by incorporating the functionally important residues from κ-hefutoxin 1 onto the Tk-AMP-X2 scaffold. The designed peptide contained the so-called essential dyad of amino acid residues significant for channel-blocking activity. Electrophysiological studies showed that although the parent peptide Tk-AMP-X2 did not present any activity against potassium channels, Tk-hefu blocked Kv1.3 channels with similar potency (IC50 ∼ 35 μm) to κ-hefutoxin 1 (IC50 ∼ 40 μm). We conclude that α-hairpinins are attractive in their simplicity as structural templates, which may be used for functional engineering and drug design.
Sigma-Aldrich
ISOGRO®-D Powder -Growth Medium, 97-99 atom % D
Sigma-Aldrich
ISOGRO®-15N Powder -Growth Medium, 98 atom % 15N
Sigma-Aldrich
ISOGRO®-15N,D Powder -Growth Medium, 98 atom % 15N, 97 atom % D
Sigma-Aldrich
ISOGRO®-13C,15N Powder -Growth Medium, 98 atom % 15N, 99 atom % 13C
Sigma-Aldrich
ISOGRO®-13C,15N,D Powder -Growth Medium, 98 atom % 15N, 97-99 atom % D, 99 atom % 13C
Sigma-Aldrich
ISOGRO®-13C Powder -Growth Medium, 99 atom % 13C