Merck
  • Oligonol, a low-molecular-weight polyphenol derived from lychee fruit, attenuates diabetes-induced renal damage through the advanced glycation end product-related pathway in db/db mice.
Oligonol, a low-molecular-weight polyphenol derived from lychee fruit, attenuates diabetes-induced renal damage through the advanced glycation end product-related pathway in db/db mice.
The Journal of nutrition (2014-06-13)
Chan Hum Park, Takako Yokozawa, Jeong Sook Noh
ABSTRACT
This study was conducted to examine whether oligonol, a low-molecular-weight polyphenol derived from lychee fruit, has an ameliorative effect on diabetes-induced alterations, such as advanced glycation end product (AGE) formation or apoptosis in the kidneys of db/db mice with type 2 diabetes. Oligonol [10 or 20 mg/(kg body weight · d), orally] was administered every day for 8 wk to prediabetic db/db mice, and its effect was compared with vehicle-treated db/db and normal control mice (m/m). The administration of oligonol decreased the elevated renal glucose concentrations and reactive oxygen species in db/db mice (P < 0.05). The increased serum urea nitrogen and creatinine concentrations, which reflect renal dysfunction in db/db mice, were substantially lowered by oligonol. Oligonol reduced renal protein expression of NAD(P)H oxidase subunits (p22 phagocytic oxidase and NAD(P)H oxidase-4), AGEs (except for pentosidine), and c-Jun N-terminal kinase B-targeting proinflammatory tumor necrosis factor-α (P < 0.05). Oligonol improved the expressions of antiapoptotic [B-cell lymphoma protein 2 (Bcl-2) and survivin] and proapoptotic [Bcl-2-associated X protein, cytochrome c, and caspase-3] proteins in the kidneys of db/db mice (P < 0.05). In conclusion, these results provide important evidence that oligonol exhibits a pleiotropic effect on AGE formation and apoptosis-related variables, representing renoprotective effects against the development of diabetic complications in db/db mice with type 2 diabetes.
Product Description
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
(+)-Catechin, analytical standard
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard
Epicatechin, primary reference standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
(−)-Epicatechin, ≥98% (HPLC), from green tea
Sigma-Aldrich
(−)-Epicatechin, ≥90% (HPLC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Supelco
(−)-Epicatechin, analytical standard