Merck
  • Possible allostery and oligomerization of recombinant plastidial sn-glycerol-3-phosphate acyltransferase.
Possible allostery and oligomerization of recombinant plastidial sn-glycerol-3-phosphate acyltransferase.
Archives of biochemistry and biophysics (2014-05-21)
Xue Chen, Robin Miles, Crystal Snyder, Martin Truksa, Jian Zhang, Saleh Shah, Randall J Weselake
ABSTRACT
Plastidial acyl-acyl carrier protein:sn-glycerol-3-phosphate acyltransferase (GPAT; EC 2.3.1.15) catalyzes the acyl-acyl carrier protein-dependent sn-1 acylation of sn-glycerol 3-phosphate (G3P) to produce lysophosphatic acid. Functional recombinant Erysimum asperum GPAT (EaGPAT), devoid of transit peptide, was produced in yeast. Analysis of the dependence of EaGPAT activity on increasing G3P concentration resulted in a hyperbolic response. EaGPAT exhibited a preference for 18-carbon unsaturated acyl-CoAs. Assays with concentrations of oleoyl-CoA up to 90μM revealed an exponential response to increasing concentrations of acyl donor, and the introduction of increasing concentrations of unlabeled linoleoyl-CoA into the standard reaction mixture resulted in increased incorporation of radiolabeled oleoyl moieties into lysophosphatidic acid. Collectively, the kinetic results suggest that acyl-CoA may act as both substrate and allosteric effector. EaGPAT was also shown to oligomerize to form higher molecular mass multimers, with the monomer and trimer being the predominant forms of the enzyme. Since most allosteric enzyme exhibit quaternary structure, the self-associating properties of EaGPAT are consistent with those of an allosteric enzyme. These results could have important regulatory implications when plastidial GPAT is introduced into a cytoplasmic environment where acyl-CoA is the acyl donor supporting cytoplasmic glycerolipid assembly.
Sigma-Aldrich
sn-Glycerol 3-phosphate lithium salt, ≥95.0% (TLC)
Sigma-Aldrich
DL-α-Glycerol phosphate magnesium salt hydrate, ~85% (KT)