Merck
  • Synthesis and pharmacological screening of certain N-substituted amides structurally related to some local anesthetics.
Synthesis and pharmacological screening of certain N-substituted amides structurally related to some local anesthetics.
Die Pharmazie (1979-01-01)
F El-Zahraa, S El-Basil, M El-Sayed, K M Ghoneim, M Khalifa
PMID432249
ABSTRACT
Diphenylacetyl chloride and pivaloyl chloride have been condensed with a wide variety of amines. Some of the resulting amides showed local anesthetic properties higher than those of procaine hydrochloride.