Merck
  • Pharmacokinetic interactions with felbamate. In vitro-in vivo correlation.
Pharmacokinetic interactions with felbamate. In vitro-in vivo correlation.
Clinical pharmacokinetics (1997-10-07)
P Glue, C R Banfield, J L Perhach, G G Mather, J K Racha, R H Levy
ABSTRACT
This article provides an analysis of the degree of agreement between in vivo interaction studies performed in patients with epilepsy and healthy individuals, and in vitro studies which identified the cytochromes P450 (CYP) inhibited by felbamate and those involved in its metabolism. In vitro studies show that felbamate is a substrate for CYP3A4 and CYP2E1. Compounds which induce CYP3A4 (e.g. carbamazepine, phenytoin and phenobarbital) increase felbamate clearance. However, the CYP3A4 inhibitors gestodene, ethinyl estradiol and erythromycin have little or no effect on felbamate trough plasma concentrations, consistent with the fact that the pathway is relatively minor for felbamate under normal (non-induced) conditions. Felbamate has been shown in vitro to inhibit CYP2C19, which would account for its effect on phenytoin clearance, and it had been postulated that this could be the mechanism underlying the reduced clearance of phenobarbital by felbamate. Although not yet examined in vitro, felbamate appears to induce the activity of CYP3A4, which would account for it reducing plasma concentrations of carbamazepine or the progestin gestodene. Interactions involving felbamate and non-CYP450-mediated metabolic pathways have also been addressed in clinical studies. The reduction in valproic acid (valproate sodium) clearance by felbamate is through the inhibition of beta-oxidation. No clinically relevant pharmacokinetic interactions were noted between felbamate and lamotrigine, clonazepam, vigabatrin, nor the active monohydroxy metabolite of oxcarbazepine. Information on the mechanisms underlying felbamate's drug:drug interaction profile permits predictions to be made concerning the likelihood of interactions with other compounds.
Sigma-Aldrich
Felbamate
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
1 Available to ship on 20 December 2024
Details...
€206.00
50 mg
1 Available to ship on 20 December 2024
Details...
€705.00
€493.50