Merck
  • Acarbose: oral anti-diabetes drug with additional cardiovascular benefits.
Acarbose: oral anti-diabetes drug with additional cardiovascular benefits.
Expert review of cardiovascular therapy (2008-02-06)
Markolf Hanefeld, Frank Schaper
ABSTRACT
Acarbose is an alpha-glucosidase inhibitor acting specifically at the level of postprandial glucose excursion. This compound lowers HbA(1c) by 0.5-1% in patients with Type 2 diabetes, either drug naive or in combination with other antidiabetic drugs. In those with impaired glucose tolerance (IGT), it reduces the incidence of newly diagnosed diabetes by 36.4%. Furthermore, it has beneficial effects on overweight, reduces blood pressure and triglycerides, and downregulates biomarkers of low-grade inflammation. In the Study To Prevent Non-Insulin-Dependent-Diabetes-Mellitus (STOP-NIDDM) trial, acarbose significantly reduced the progression of intima media thickness, incidence of cardiovascular events and of newly diagnosed hypertension. In a meta-analysis of patients with Type 2 diabetes (MERIA), acarbose intake was associated with a reduction of cardiovascular events by 35%. Acarbose is a very safe drug but in approximately 30% of patients, it can cause gastrointestinal complaints due to its mode of action, which in the majority disappear after 1-2 months. Acarbose is approved for treatment of IGT in 25 countries. It can be given alone or in combination with other oral antidiabetics and insulin. Acarbose is particularly effective in those with IGT and early diabetes and patients with comorbidities of the metabolic syndrome.
Product Description
Acarbose for identification, European Pharmacopoeia (EP) Reference Standard
Acarbose for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acarbose, ≥95% (HPLC)
Acarbose, European Pharmacopoeia (EP) Reference Standard
Supelco
Acarbose, Pharmaceutical Secondary Standard; Certified Reference Material