Merck
Role of non-protein amino acid L-canavanine in autoimmunity.
Autoimmunity reviews (2006-08-08)
Jun Akaogi, Tolga Barker, Yoshiki Kuroda, Dina C Nacionales, Yoshioki Yamasaki, Bruce R Stevens, Westley H Reeves, Minoru Satoh
ABSTRACT
Association of SLE and alfalfa was first reported in a volunteer who developed lupus-like autoimmunity while ingesting alfalfa seed for a hypercholesterolemia study. This was corroborated with studies in monkeys fed with alfalfa sprout that developed SLE. Re-challenge with L-canavanine relapsed the disease. Arginine homologue L-canavanine, present in alfalfa, was suspected as a cause. L-canavanine can be charged by arginyl tRNA synthetase to replace L-arginine during protein synthesis. Aberrant canavanyl proteins have disrupted structure and functions. Induction or exacerbation of SLE by alfalfa tablets reported in a few cases remains controversial. Epidemiological studies on the relationship between alfalfa and SLE are sparse. In mice, NZB/W F1, NZB, and DBA/2 mice fed with L-canavanine show exacerbation/triggering of the SLE, however, BALB/c studies were negative. L-canavanine incorporation may be more efficient in the presence of inflammation or other conditions that can cause arginine deficiency. The L-canavanine induced apoptotic cells can be phagocytosed and a source of autoantigens processed by endosomal proteases. Endogenous canavanyl proteins are ubiquitinated and processed via proteasome. Incorporation of L-canavanine into proteasome or endosome can also cause disruption of antigen processing. Alfalfa/L-canavanine-induced lupus will be an interesting model of autoimmunity induced by the modification of self-proteins at the translational level.
Sigma-Aldrich
L-Canavanine sulfate salt, ≥99% (TLC), powder
Sigma-Aldrich
L-Canavanine, ≥98% (TLC), powder, from Canavalia ensiformis