Merck
Does 2,3-butanedione monoxime inhibit nonmuscle myosin?
Protoplasma (2005-05-04)
Arthur Forer, Lacramioara Fabian
ABSTRACT
BDM (2,3-butanedione monoxime) has been used extensively to inhibit nonmuscle myosin. However, recent articles raise the question of what BDM actually does, because of experiments in which BDM did not affect the actin-activated ATPase of nonmuscle myosins. We describe results that indicate that BDM indeed inhibits motility due to nonmuscle myosins: in many different cells BDM has the same effects as anti-actin agents and/or as other anti-myosin agents, and BDM slows or stops the sliding between actin filaments and myosin in vitro. We discuss how the two sets of apparently contradictory results might be resolved, and we suggest possible experiments that might clarify the contradictory interpretations.
Sigma-Aldrich
2,3-Butanedione monoxime, ≥98%
Supelco
2,3-Butanedione monoxime, for spectrophotometric det. of urea, ≥99.0%
Sigma-Aldrich
2,3-Butanedione, 97%
Supelco
2,3-Butanedione, analytical standard