MilliporeSigma
  • Resveratrol analogues as selective cyclooxygenase-2 inhibitors: synthesis and structure-activity relationship.
Resveratrol analogues as selective cyclooxygenase-2 inhibitors: synthesis and structure-activity relationship.
Bioorganic & medicinal chemistry (2004-10-07)
Marek Murias, Norbert Handler, Thomas Erker, Karin Pleban, Gerhard Ecker, Philipp Saiko, Thomas Szekeres, Walter Jäger
ABSTRACT
Resveratrol (3,5,4'-trihydroxy-trans-stilbene) is found in grapes and various medical plants. Among cytotoxic, antifungal, antibacterial cardioprotective activity resveratrol also demonstrates non-selective cyclooxygenase-1 (COX-1) and cyclooxygenase-2 (COX-2) inhibition. In order to find more selective COX-2 inhibitors a series of methoxylated and hydroxylated resveratrol derivatives were synthesized and evaluated for their ability to inhibit both enzymes using in vitro inhibition assays for COX-1 and COX-2 by measuring PGE(2) production. Hydroxylated but not methoxylated resveratrol derivatives showed a high rate of inhibition. The most potent resveratrol compounds were 3,3',4',5-tetra-trans-hydroxystilbene (COX-1: IC(50)=4.713, COX-2: IC(50)=0.0113 microM, selectivity index=417.08) and 3,3',4,4',5,5'-hexa-hydroxy-trans-stilbene (COX-1: IC(50)=0.748, COX-2: IC(50)=0.00104 microM, selectivity index=719.23). Their selectivity index was in part higher than celecoxib, a selective COX-2 inhibitor already established on the market (COX-1: IC(50)=19.026, COX-2: IC(50)=0.03482 microM, selectivity index=546.41). Effect of structural parameters on COX-2 inhibition was evaluated by quantitative structure-activity relationship (QSAR) analysis and a high correlation was found with the topological surface area TPSA (r=0.93). Docking studies on both COX-1 and COX-2 protein structures also revealed that hydroxylated but not methoxylated resveratrol analogues are able to bind to the previously identified binding sites of the enzymes. Hydroxylated resveratrol analogues therefore represent a novel class of highly selective COX-2 inhibitors and promising candidates for in vivo studies.