Merck
  • Reduced prevalence of Giardia duodenalis in iron-deficient Rwandan children.
Reduced prevalence of Giardia duodenalis in iron-deficient Rwandan children.
Tropical medicine & international health : TM & IH (2014-06-06)
Ina Danquah, Jean-Bosco Gahutu, Ralf Ignatius, Andre Musemakweri, Frank P Mockenhaupt
ABSTRACT
Acute symptomatic infection with Giardia duodenalis impairs iron absorption, but iron deficiency may protect against infections caused by various micro-organisms including parasites. We therefore examined the association of G. duodenalis infection and iron deficiency in 575 Rwandan children under 5 years of age. Giardia duodenalis infection was diagnosed by triplicate microscopy and PCR assays, and iron deficiency was defined as a ferritin concentration <12 ng/ml. Largely asymptomatic G. duodenalis infection was seen in 65.3% of the children and iron deficiency in 17.4%. G. duodenalis infection was less common in iron-deficient children (51%) than in non-deficient children (68%, P = 0.002). In multivariate analysis, the odds of G. duodenalis infection were almost halved in iron-deficient children (adjusted odds ratio, 0.54; 95% confidence interval, 0.33-0.86). In this highly endemic setting, there was no evidence that Giardia infection impairs iron status. Rather, iron deficiency appeared to protect against infection with this parasite.
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Iron, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Iron, powder, −325 mesh, 97%
Sigma-Aldrich
C Reactive Protein from human fluids, buffered aqueous solution
Sigma-Aldrich
Ferritin from equine spleen, Type I, saline solution
Sigma-Aldrich
Ferritin from human spleen, Type V, 10 μg/mL in 0.15 M NaCl, 10 mM Tris, pH 8.0, containing 0.1% sodium azide
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Iron, foil, 2m coil, thickness 0.038mm, hard, 99.5%
Iron, foil, 4mm disks, thickness 0.5mm, hard, 99.5%
Iron, foil, 4mm disks, thickness 0.125mm, as rolled, 99.99+%
Iron, foil, 25mm disks, thickness 0.015mm, 99.85%
Iron, foil, 4mm disks, thickness 0.0125mm, 99.99+%
Iron, foil, 25mm disks, thickness 0.025mm, hard, 99.5%
Iron, foil, 4mm disks, thickness 0.9mm, as rolled, 99.5%
Iron, foil, light tested, 25x25mm, thickness 0.015mm, 99.85%
Iron, foil, 15mm disks, thickness 0.003mm, 99.85%
Iron, foil, 10mm disks, thickness 0.025mm, as rolled, 99.99+%
Iron, foil, 1m coil, thickness 0.5mm, hard, 99.5%
Iron, foil, 10mm disks, thickness 0.25mm, as rolled, 99.99+%
Iron, foil, 20m coil, thickness 0.038mm, hard, 99.5%
Iron, foil, 1m coil, thickness 0.075mm, hard, 99.5%
Iron, foil, 15mm disks, thickness 0.006mm, 99.85%
Iron, foil, 15mm disks, thickness 0.25mm, hard, 99.5%
Iron, foil, 15mm disks, thickness 0.020mm, 99.85%
Iron, foil, 10mm disks, thickness 0.38mm, hard, 99.5%
Iron, foil, 10mm disks, thickness 0.075mm, as rolled, 99.99+%
Iron, foil, 20m coil, thickness 0.075mm, hard, 99.5%
Iron, foil, 15mm disks, thickness 0.38mm, hard, 99.5%
Iron, foil, 25mm disks, thickness 0.075mm, hard, 99.5%