Merck
  • Steroids in aminoglycoside-containing ear drops: do they reduce cochlear toxicity?
Steroids in aminoglycoside-containing ear drops: do they reduce cochlear toxicity?
The Laryngoscope (2013-10-22)
Brian W Blakley, Saad Alsaleh, Zameel Dewji, Kaiser Qureshy, Sylvie Berard, Ling Xie
ABSTRACT
To determine whether betamethasone (BM) reduces the cochlear toxicity of otic gentamicin (GM) if given together. Controlled animal study. Thirty-four mice were assigned at random to receive intratympanic injections of either 0.1 % BM (11 mice), 0.3% GM (13 mice), or a combination of both (GM/BM) with benzalkonium chloride (10 mice) in the left ear (treated) and saline on the right (untreated). Six injections were given on alternate days. Auditory brainstem response thresholds were assessed at 1 month, 2 months, and >2 months. There was a significantly greater degree of hearing loss in the BM-treated ears compared to the untreated ears (6.48 dB hearing loss, P = .007) and in the GM-treated ears compared to untreated ears (6.59 dB hearing loss, P = .010,). However, otic GM/BM and benzalkonium chloride did not cause significant additional hearing loss compared with the untreated ears (3.56 dB hearing loss, P = .242). Our data suggest that hearing loss caused by GM otic drops may be reduced by the inclusion of BM and benzalkonium chloride. Our finding that BM alone was associated with hearing loss suggests that the benzalkonium chloride may be the protective agent in combination otic drops.
Product Description
Supelco
Betamethasone, VETRANAL®, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Betamethasone, ≥98%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Betamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Betamethasone, European Pharmacopoeia (EP) Reference Standard