Merck
  • Auxin distribution is differentially affected by nitrate in roots of two rice cultivars differing in responsiveness to nitrogen.
Auxin distribution is differentially affected by nitrate in roots of two rice cultivars differing in responsiveness to nitrogen.
Annals of botany (2013-10-08)
Wenjing Song, Huwei Sun, Jiao Li, Xianpo Gong, Shuangjie Huang, Xudong Zhu, Yali Zhang, Guohua Xu
ABSTRACT
Although ammonium (NH4(+)) is the preferred form of nitrogen over nitrate (NO3(-)) for rice (Oryza sativa), lateral root (LR) growth in roots is enhanced by partial NO3(-) nutrition (PNN). The roles of auxin distribution and polar transport in LR formation in response to localized NO3(-) availability are not known. Time-course studies in a split-root experimental system were used to investigate LR development patterns, auxin distribution, polar auxin transport and expression of auxin transporter genes in LR zones in response to localized PNN in 'Nanguang' and 'Elio' rice cultivars, which show high and low responsiveness to NO3(-), respectively. Patterns of auxin distribution and the effects of polar auxin transport inhibitors were also examined in DR5::GUS transgenic plants. Initiation of LRs was enhanced by PNN after 7 d cultivation in 'Nanguang' but not in 'Elio'. Auxin concentration in the roots of 'Nanguang' increased by approx. 24 % after 5 d cultivation with PNN compared with NH4(+) as the sole nitrogen source, but no difference was observed in 'Elio'. More auxin flux into the LR zone in 'Nanguang' roots was observed in response to NO3(-) compared with NH4(+) treatment. A greater number of auxin influx and efflux transporter genes showed increased expression in the LR zone in response to PNN in 'Nanguang' than in 'Elio'. The results indicate that higher NO3(-) responsiveness is associated with greater auxin accumulation in the LR zone and is strongly related to a higher rate of LR initiation in the cultivar 'Nanguang'.
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-3, aqueous solution, ≥90,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-3AF, aqueous solution, ≥60,000 units/mL
Sigma-Aldrich
β-Glucuronidase from limpets (Patella vulgata), Type L-II, lyophilized powder, 1,000,000-3,000,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type HP-2, aqueous solution, ≥100,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-2, aqueous solution, ≥85,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-1, partially purified powder, ≥300,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type HP-2S, aqueous solution, ≥90,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-5, lyophilized powder, ≥400,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from limpets (Patella vulgata), aqueous solution, ≥85,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, ≥10,000,000 units/g protein (30 min assay), recombinant, expressed in E. coli overproducing strain, lyophilized powder
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, ≥20,000 units/mg protein, recombinant, expressed in E. coli overproducing strain, lyophilized powder
Sigma-Aldrich
β-Glucuronidase from Helix aspersa (garden snail), Type HA-4
Sigma-Aldrich
β-Glucuronidase from bovine liver, Type B-3, ≥2,000,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, Type IX-A, lyophilized powder, 1,000,000-5,000,000 units/g protein (30 min assay)
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, aqueous glycerol solution, ≥5,000,000 units/g protein, pH 6.8 (biuret)
Sigma-Aldrich
β-Glucuronidase from bovine liver, Type B-1, ≥1,000,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, Type VII-A, lyophilized powder, 5,000,000-20,000,000 units/g protein, pH 6.8 (30 min assay)
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, >20,000,000 units/g protein, recombinant, expressed in E. coli, aqueous glycerol solution