Merck
  • Scalable purification of the lantibiotic nisin and isolation of chemical/enzymatic cleavage fragments suitable for semi-synthesis.
Scalable purification of the lantibiotic nisin and isolation of chemical/enzymatic cleavage fragments suitable for semi-synthesis.
Journal of peptide science : an official publication of the European Peptide Society (2013-09-12)
Jack C Slootweg, Rob M J Liskamp, Dirk T S Rijkers
ABSTRACT
Herein, we describe a scalable purification of the lantibiotic nisin via an extraction/precipitation approach using a biphasic system, which can be carried out up to 40-80 gram scale. This approach results in an at least tenfold enrichment of commercially available preparations of nisin, which usually contain only 2.5% of the desired peptide, to allow further purification by preparative HPLC. As a follow-up study, the enriched nisin sample was digested either by trypsin or chymotrypsin, or treated by CNBr, and these reactions were monitored by LC-MS to identify and characterize the obtained fragments. Two previously unknown cleavage sites have been identified: Asn20-Met21 and Met21-Lys22 for trypsin and chymotrypsin, respectively. Furthermore, a novel and convenient enzymatic approach to isolate the native nisin C-ring [nisin fragment (13-20)] was uncovered. Finally, by means of preparative HPLC, nisin fragments (1-12), (1-20), (22-34), and (22-31) could be isolated and will be used in a semi-synthesis approach to elucidate the role of each fragment in the mode of action of nisin as an antimicrobial peptide.
Sigma-Aldrich
α-Chymotrypsin−Agarose from bovine pancreas, lyophilized powder, 2,000-3,500 units/g agarose (One ml gel will yield 65-120 units)
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, Type II, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, ≥40 units/mg protein, vial of 5 mg
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, Type I-S, essentially salt-free, lyophilized powder
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, suitable for protein sequencing, salt-free, lyophilized powder
Sigma-Aldrich
α-Chymotrypsin from human pancreas, lyophilized powder
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, (TLCK treated to inactivate residual tryspin activity), Type VII, essentially salt-free, lyophilized powder, ≥40 units/mg protein