Merck
  • Characterization of deamidation at Asn138 in L-chain of recombinant humanized Fab expressed from Pichia pastoris.
Characterization of deamidation at Asn138 in L-chain of recombinant humanized Fab expressed from Pichia pastoris.
Journal of biochemistry (2013-07-16)
Takatoshi Ohkuri, Eri Murase, Shu-Lan Sun, Jun Sugitani, Tadashi Ueda
ABSTRACT
A method was previously established for evaluating Asn deamidation by matrix-assisted laser desorption/ionization time of flight-mass spectrometry using endoproteinase Asp-N. In this study, we demonstrated that this method could be applied to the identification of the deamidation site of the humanized fragment antigen-binding (Fab). First, a system for expressing humanized Fab from methylotrophic yeast Pichia pastoris was constructed, resulting in the preparation of ∼30 mg of the purified humanized Fab from 1 l culture. Analysis of the L-chain derived from recombinant humanized Fab that was heated at pH 7 and 100°C for 1 h showed the deamidation at Asn138 in the constant region. Then, we prepared L-N138D Fab and L-N138A Fab and examined their properties. The circular dichroism (CD) spectrum of the L-N138D Fab was partially different from that of the wild-type Fab. The measurement of the thermostability showed that L-N138D caused a significant decrease in the thermostability of Fab. On the other hand, the CD spectrum and thermostability of L-N138A Fab showed the same behaviour as the wild-type Fab. Thus, it was suggested that the introduction of a negative charge at position 138 in the L-chain by the deamidation significantly affected the stability of humanized Fab.
Sigma-Aldrich
Endoproteinase Asp-N from Pseudomonas fragi mutant strain, suitable for protein sequencing, lyophilized powder