Merck
  • Identification and determination of ketotifen hydrogen fumarate, azelastine hydrochloride, dimetindene maleate and promethazine hydrochloride by densitometric method.
Identification and determination of ketotifen hydrogen fumarate, azelastine hydrochloride, dimetindene maleate and promethazine hydrochloride by densitometric method.
Acta poloniae pharmaceutica (2014-01-05)
Elzbieta Wyszomirska, Krystyna Czerwińska, Elzbieta Kublin, Aleksander P Mazurek
ABSTRACT
Conditions for determination of: ketotifen hydrogen fumarate, azelastine hydrochloride, dimetindene maleate and promethazine hydrochloride by densitometric method in substances and pharmaceuticals were provided. Maximum wavelenghts were: 228 nm for ketotifen hydrogen fumarate, 295 nm for azelastine hydrochloride, 265 nm for dimetindene maleate and 255 nm for promethazine hydrochloride. The limits of quantification were in the ranges of 0.2-5 microg/spot. The statistical data showed adequate accuracy and precision of developed methods.
Sigma-Aldrich
Ketotifen fumarate salt
Pricing and availability is not currently available.
Dimetindene maleate, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Ketotifen for peak identification, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Ketotifen hydrogen fumarate, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.