Merck
  • Efficacy of rivastigmine in comparison to ginkgo for treating Alzheimer's dementia.
Efficacy of rivastigmine in comparison to ginkgo for treating Alzheimer's dementia.
JPMA. The Journal of the Pakistan Medical Association (2013-07-23)
Nastaran Majdi Nasab, Mohammad Ali Bahrammi, Mohammad Reza Amiri Nikpour, Fakher Rahim, Sayed Navid Naghibis
ABSTRACT
To assess the efficacy of the Ginkgo biloba in patients with dementia of the Alzheimer type in slowing down the disease's degenerative progression and the patients' cognitive impairment compared with rivastigmine. Total 56 patients aged 50-75 years, suffering from dementia, were allocated into one of the two treatments: group 1) Ginkgo biloba (120 mg daily dose); group 2) rivastigmine (4.5 mg daily dose) in a 24-week randomized double blind study. The degree of severity of dementia was assessed by the Seven Minute test and the Mini-Mental State Examination. Our results confirm the clinical efficacy of rivastigmine in the dementia of the Alzheimer type, comparing to Ginkgo biloba. There are few published trials that have directly compared a cholinesterase inhibitor with Ginkgo for dementia. This study directly compares a cholinesterase inhibitor with Ginkgo biloba for dementia of the Alzheimer type. Our study suggests that there are differences in the efficacy of Ginkgo biloba and rivastigmine in the treatment of Alzheimer's dementia. In addition, this study suggested that cholinesterase inhibitors should be used in preference to Ginkgo biloba in patients with mild to moderate AD.
Product Description
Rivastigmine hydrogen tartrate, European Pharmacopoeia (EP) Reference Standard
Rivastigmine for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Rivastigmine tartrate, ≥98% (HPLC)