Merck
[Rapid analysis of suppositories by quantitative 1H NMR spectroscopy].
Antibiotiki i khimioterapiia = Antibiotics and chemoterapy [sic] (2012-11-20)
R A Abramovich, S A Kovaleva, S V Goriainov, A N Vorob'ev, G A Kalabin
ABSTRACT
Rapid analysis of suppositories with ibuprofen and arbidol by quantitative 1H NMR spectroscopy was performed. Optimal conditions for the analysis were developed. The results are useful for design of rapid methods for quality control of suppositories with different components
Sigma-Aldrich
Arbidol hydrochloride, ≥98% (HPLC)