MilliporeSigma
  • Effect of decursinol angelate on the pharmacokinetics of theophylline and its metabolites in rats.
Effect of decursinol angelate on the pharmacokinetics of theophylline and its metabolites in rats.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2012-07-10)
Jung-woo Chae, Jung-hwa An, Wonku Kang, Jin yeul Ma, Kwang-il Kwon
ABSTRACT
Herb-drug interactions represent a serious problem as herbal medicine is used extensively in the modern world. This study investigated the effects of decursinol angelate on the pharmacokinetics of theophylline, a typical substrate of the cytochrome P450 1A2 enzyme, in rats. After 3 days of decursinol angelate pretreatment, on the fourth day, rats were administered decursinol angelate and theophylline concomitantly. Blood theophylline and its major metabolite [1-methylxanthine (1-MX), 3-methylxanthine (3-MX), 1-methyluric acid (1-MU), and 1,3-dimethyluric acid (1,3-DMU)] levels were monitored by liquid chromatography-tandem mass spectroscopy. The results indicated that theophylline clearance significantly decreased and the area under the concentration-time curve (AUC) increased in decursinol angelate (25 mg/kg)-pretreated rats administered theophylline (10 mg/kg). The elimination half-life (t1/2) of theophylline was increased by 20%. In the presence of decursinol angelate (25 mg/kg), the pharmacokinetic parameters of three metabolites (1-MX, 1,3-DMU, and 1-MU) were significantly altered (half-life for 1-MU, and AUC24 h for 1-MX, 1,3-DMU, and 1-MU). Our results suggest that patients receiving CYP1A2-metabolized drugs, such as caffeine and theophylline, should be advised of the potential herb-drug interaction to reduce the risk of therapeutic failure or increased toxicity of conventional drug therapy.
Sigma-Aldrich
Decursin, ≥97% (HPLC)