Merck
  • Dissolution profile of novel composite pellet cores based on different ratios of microcrystalline cellulose and isomalt.
Dissolution profile of novel composite pellet cores based on different ratios of microcrystalline cellulose and isomalt.
Journal of pharmaceutical sciences (2012-05-11)
Oliver Luhn, Nikolett Kállai, Zsombor Kristóf Nagy, Kristóf Kovács, Bodo Fritzsching, Imre Klebovich, István Antal
ABSTRACT
There is a growing interest towards the application of inert cores as starting materials for pharmaceutical pellet manufacturing. They serve as alternatives to develop and adapt a relatively simple manufacturing technology compared with an extrusion/spheronisation process. The major objective of this study was to investigate the effect of the compositions of core materials on the drug release profile. Pure microcrystalline cellulose (MCC), isomalt and different types of novel composite MCC-isomalt cores were layered with model drug (sodium diclofenac) and were coated with acrylic polymer. The effect of the osmolality in the gastrointestinal tract was simulated using glucose as osmotically active agent during in vitro dissolution tests. The results demonstrated the dependence of drug dissolution profile on the ratio of MCC and isomalt in the core and the influence of osmotic properties of the dissolution medium. Isomalt used in the composite core was able to decrease the vulnerability of the dissolution kinetics to the changes in the osmotic environment.
Product Description
Isomalt, European Pharmacopoeia (EP) Reference Standard
USP
Isomalt, United States Pharmacopeia (USP) Reference Standard