Merck
  • Postmortem whole-body CT angiography: evaluation of two contrast media solutions.
Postmortem whole-body CT angiography: evaluation of two contrast media solutions.
AJR. American journal of roentgenology (2008-04-24)
Steffen Ross, Danny Spendlove, Stephan Bolliger, Andreas Christe, Lars Oesterhelweg, Silke Grabherr, Michael J Thali, Erich Gygax
ABSTRACT
The objective of our study was to establish a standardized procedure for postmortem whole-body CT-based angiography with lipophilic and hydrophilic contrast media solutions and to compare the results of these two methods. Minimally invasive postmortem CT angiography was performed on 10 human cadavers via access to the femoral blood vessels. Separate perfusion of the arterial and venous systems was established with a modified heart-lung machine using a mixture of an oily contrast medium and paraffin (five cases) and a mixture of a water-soluble contrast medium with polyethylene glycol (PEG) 200 in the other five cases. Imaging was executed with an MDCT scanner. The minimally invasive femoral approach to the vascular system provided a good depiction of lesions of the complete vascular system down to the level of the small supplying vessels. Because of the enhancement of well-vascularized tissues, angiography with the PEG-mixed contrast medium allowed the detection of tissue lesions and the depiction of vascular abnormalities such as pulmonary embolisms or ruptures of the vessel wall. The angiographic method with a water-soluble contrast medium and PEG as a contrast-agent dissolver showed a clearly superior quality due to the lack of extravasation through the gastrointestinal vascular bed and the enhancement of soft tissues (cerebral cortex, myocardium, and parenchymal abdominal organs). The diagnostic possibilities of these findings in cases of antemortem ischemia of these tissues are not yet fully understood.
Product Description
USP
Iodixanol Related Compound D, United States Pharmacopeia (USP) Reference Standard
Iopentol, European Pharmacopoeia (EP) Reference Standard