Merck
  • In vitro studies on the genotoxicity of the organophosphorus insecticide malathion and its two analogues.
In vitro studies on the genotoxicity of the organophosphorus insecticide malathion and its two analogues.
Mutation research (1999-11-27)
J Błasiak, P Jałoszynski, A Trzeciak, K Szyfter
ABSTRACT
Malathion [S-(1,2-dicarboethoxyethyl)O,O-dimethyl phosphorodithioate] is a commonly used organophosphorus insecticide reported to be genotoxic both in vivo and in vitro, but the reports are conflicting. In order to elucidate the genotoxic potency of the main compounds present in commercial preparations of malathion, the DNA-damaging effect of this insecticide, its major metabolite malaoxon [S-(1,2-dicarboethoxyethyl)O,O-dimethyl phosphorothiolate] and its isomer isomalathion [S-(1,2-dicarboethoxyethyl)O,S-dimethyl phosphorodithioate], all at purity of at least 99.8%, was investigated by use of the alkaline single cell gel electrophoresis (comet assay). Freshly isolated human peripheral blood lymphocytes were incubated with 25, 75 and 200 microM of the chemicals for 1 h at 37 degrees C. The concentrations used are comparable to those found in blood following various non-lethal human exposures to pesticides. Malathion did not cause any significant changes in the comet length of the lymphocytes, throughout the range of concentrations tested. Malaoxon and isomalathion introduced damage to DNA in a dose-dependent manner. The effect induced by malaoxon was more pronounced than that caused by isomalathion. Treated cells were able to recover within a 60-min incubation in insecticide-free medium at 37 degrees C except the lymphocytes exposed to malaoxon at 200 microM, which did not show measurable DNA repair. The latter result suggests a considerable cytotoxic effect (cell death) of malaoxon at the highest concentration used. The reported genotoxicity of malathion might, therefore, be a consequence of its metabolic biotransformation to malaoxon or the presence of malaoxon and/or isomalathion as well as other unspecified impurities in commercial formulations of malathion. In this regard, the results of our study clearly indicate that malathion used as commercial product, i.e., containing malaoxon and isomalathion, can be considered as a genotoxic substance in vitro. This means that it may also produce DNA disturbances in vivo, such as DNA breakage at sites of oncogenes or tumor suppressor genes, thus playing a role in the induction of malignancies in individuals exposed to this agent. Therefore, malathion can be regarded as a potential mutagen/carcinogen and requires further investigation.
Product Description
Malathion impurity A, European Pharmacopoeia (EP) Reference Standard