Merck
  • Double nucleophilic attack on isocyanide carbon: a synthetic strategy for 7-aza-tetrahydroindoles.
Double nucleophilic attack on isocyanide carbon: a synthetic strategy for 7-aza-tetrahydroindoles.
Chemical communications (Cambridge, England) (2012-11-13)
Yifei Li, Xianxiu Xu, Chunyu Xia, Lingjuan Zhang, Ling Pan, Qun Liu
ABSTRACT
A novel and efficient route for the synthesis of 7-aza-tetrahydroindoles from N-aryl/alkyl-alkenoylacetamides and ethyl isocyanoacetate is described. A mechanism, involving a stepwise [3+2] cycloaddition-intramolecular aza-Michael addition cascade, is proposed that explains the origin of the double nucleophilic attack on the isocyanide carbon atom.
Sigma-Aldrich
Ethyl isocyanoacetate, 95%