Merck
On-line single droplet deposition for MALDI mass spectrometry.
Journal of the American Society for Mass Spectrometry (2004-10-07)
Xia Zhang, Damien A Narcisse, Kermit K Murray
ABSTRACT
A single droplet generator was coupled with a rotating ball inlet matrix-assisted laser desorption/ionization (MALDI) time of flight (TOF) mass spectrometer. Single droplets with 100 picoliter volume were ejected by a piezoelectric-actuated droplet generator and deposited onto a matrix-coated rotating stainless steel ball at atmospheric pressure. The single droplet deposit was transported to the vacuum side of the instrument where ionization was accomplished using a UV pulsed laser. Using this on-line interface, it was possible to obtain protonated molecule signal from as little as 10 fmol analyte.
Sigma-Aldrich
Cytochrome c from equine heart, ≥95% (SDS-PAGE)
Sigma-Aldrich
Angiotensin I human acetate salt hydrate, ≥90% (HPLC), powder, suitable for mammalian cell culture
Sigma-Aldrich
Myoglobin from equine heart, ≥90% (SDS-PAGE), essentially salt-free, lyophilized powder