Merck
  • The preparation of 3-substituted-1,5-dibromopentanes as precursors to heteracyclohexanes.
The preparation of 3-substituted-1,5-dibromopentanes as precursors to heteracyclohexanes.
Beilstein journal of organic chemistry (2011-04-23)
Bryan Ringstrand, Martin Oltmanns, Jeffrey A Batt, Aleksandra Jankowiak, Richard P Denicola, Piotr Kaszynski
ABSTRACT
The methodology to prepare 3-substituted 1,5-dibromopentanes I and their immediate precursors, which include 3-substituted 1,5-pentanediols VII or 4-substituted tetrahydropyrans VIII, is surveyed. Such dibromides I are important intermediates in the preparation of liquid crystalline derivatives containing 6-membered heterocyclic rings. Four dibromides 1a-1d containing simple alkyl and more complex fragments at the 3-position were prepared. 3-Propyl- and 3-pentyl-pentane-1,5-diol (2a,b) were prepared starting from either glutaconate or malonate diesters, while tetrahydropyrans 3c and 3d were obtained from tetrahydro-4H-pyran-4-one. The advantages and disadvantages of each route are discussed. Dibromides 1c and 1d were used to prepare sulfonium zwitterions 11c and 11d.
Sigma-Aldrich
Tetrahydro-4H-pyran-4-one, 99%
Pricing and availability is not currently available.