Merck
  • New synthesis of zinc tetrakis(arylethynyl)porphyrins and substituent effects on their redox chemistry.
New synthesis of zinc tetrakis(arylethynyl)porphyrins and substituent effects on their redox chemistry.
Dalton transactions (Cambridge, England : 2003) (2007-03-28)
Ming-Cheng Kuo, Long-An Li, Wei-Nan Yen, Shang-Shih Lo, Cheng-Wei Lee, Chen-Yu Yeh
ABSTRACT
Sonogashira coupling of zinc 5,10,15,20-tetraethynylporphyrin with various phenyl iodides under mild conditions afforded good yields of the corresponding zinc porphyrins. This method is applicable to a variety of aryl iodides including meso-substituted iodoporphyrin to form a conjugated star-shaped multiporphyrin. The UV-Vis spectra show that peak broadening, red shifts, and changes in the oscillator strength of absorptions increase with the extension of pi-conjugation. In the electrochemical measurements, the first oxidation of porphyrins 4-9 occurs at potentials in the range +0.89 to +1.08 V, which are comparable to that of ZnTPP (TPP = tetraphenylporphyrin). The first reduction was observed at potentials from -0.73 to -0.89 V, which is anodically shifted by 390-550 mV as compared to that of ZnTPP, and the second reduction occurs at potentials in the range -1.12 to -1.33 V. The para-substituted tetrakis(phenylethynyl)porphyrins show substituent effects on their redox chemistry and exhibit only slight substituent effects in their emission and absorption maxima.
Sigma-Aldrich
3-Iodobenzotrifluoride, 98%
Pricing and availability is not currently available.