Merck
  • mTOR target NDRG1 confers MGMT-dependent resistance to alkylating chemotherapy.
mTOR target NDRG1 confers MGMT-dependent resistance to alkylating chemotherapy.
Proceedings of the National Academy of Sciences of the United States of America (2013-12-25)
Markus Weiler, Jonas Blaes, Stefan Pusch, Felix Sahm, Marcus Czabanka, Sebastian Luger, Lukas Bunse, Gergely Solecki, Viktoria Eichwald, Manfred Jugold, Sibylle Hodecker, Matthias Osswald, Christoph Meisner, Thomas Hielscher, Petra Rübmann, Philipp-Niklas Pfenning, Michael Ronellenfitsch, Tore Kempf, Martina Schnölzer, Amir Abdollahi, Florian Lang, Martin Bendszus, Andreas von Deimling, Frank Winkler, Michael Weller, Peter Vajkoczy, Michael Platten, Wolfgang Wick
ABSTRACT
A hypoxic microenvironment induces resistance to alkylating agents by activating targets in the mammalian target of rapamycin (mTOR) pathway. The molecular mechanisms involved in this mTOR-mediated hypoxia-induced chemoresistance, however, are unclear. Here we identify the mTOR target N-myc downstream regulated gene 1 (NDRG1) as a key determinant of resistance toward alkylating chemotherapy, driven by hypoxia but also by therapeutic measures such as irradiation, corticosteroids, and chronic exposure to alkylating agents via distinct molecular routes involving hypoxia-inducible factor (HIF)-1alpha, p53, and the mTOR complex 2 (mTORC2)/serum glucocorticoid-induced protein kinase 1 (SGK1) pathway. Resistance toward alkylating chemotherapy but not radiotherapy was dependent on NDRG1 expression and activity. In posttreatment tumor tissue of patients with malignant gliomas, NDRG1 was induced and predictive of poor response to alkylating chemotherapy. On a molecular level, NDRG1 bound and stabilized methyltransferases, chiefly O(6)-methylguanine-DNA methyltransferase (MGMT), a key enzyme for resistance to alkylating agents in glioblastoma patients. In patients with glioblastoma, MGMT promoter methylation in tumor tissue was not more predictive for response to alkylating chemotherapy in patients who received concomitant corticosteroids.
Sigma-Aldrich
Hexokinase from Saccharomyces cerevisiae, lyophilized powder, ≥350 units/mg protein, Protein ≥10 % by biuret
Sigma-Aldrich
Hexokinase from Saccharomyces cerevisiae, Type F-300, lyophilized powder, ≥130 units/mg protein (biuret)