Merck
  • Pouteria ramiflora extract inhibits salivary amylolytic activity and decreases glycemic level in mice.
Pouteria ramiflora extract inhibits salivary amylolytic activity and decreases glycemic level in mice.
Anais da Academia Brasileira de Ciencias (2013-09-27)
Neire M De Gouveia, Cibele L De Albuquerque, Laila S Espindola, Foued S Espindola
ABSTRACT
In this study, extracts of plant species from the Cerrado biome were assessed in order to find potential inhibitors of human salivary alpha-amylase. The plants were collected and extracts were obtained from leaves, bark, and roots. We performed a preliminary phytochemical analysis and a screening for salivar alpha-amylase inhibitory activity. Only three botanical families (Sapotaceae, Sapindaceae and Flacourtiaceae) and 16 extracts showed a substantial inhibition (>75%) of alpha-amylase. The ethanolic extracts of Pouteria ramiflora obtained from stem barks and root barks decreased amylolytic activity above 95% at a final concentration of 20 µg/mL. Thus, adult male Swiss mice were treated orally with P. ramiflora in acute toxicity and glycemic control studies. Daily administration with 25, 50 and 100 mg/kg of aqueous extract of P. ramiflora for eight days can reduce significantly body weight and blood glucose level in mice. These data suggest that the crude polar extract of P. ramiflora decreases salivary amylolytic activity while lowering the blood levels of glucose.
Sigma-Aldrich
α-Amylase from porcine pancreas, Type VI-B, ≥5 units/mg solid
Sigma-Aldrich
Taka-Diastase from Aspergillus oryzae, powder, slightly beige, ~100 U/mg
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~380 U/mg
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~50 U/mg
Sigma-Aldrich
α-Amylase from porcine pancreas, Type I-A, PMSF treated, saline suspension, 700-1400 units/mg protein (E1%/280)
Sigma-Aldrich
α-Amylase, heat-stable, solution, for use in Total Dietary Fiber Assay, TDF-100A
Sigma-Aldrich
α-Amylase from Bacillus sp., Type II-A, lyophilized powder, ≥1,500 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, ≥400 units/mg protein (Lowry)
Sigma-Aldrich
α-Amylase from porcine pancreas, PMSF Treated, Type I-A, saline suspension, ≥1000 units/mg protein (E1%/280)
Sigma-Aldrich
α-Amylase from Bacillus sp., liquid
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~1.5 U/mg (~0.2 U acc. to Willstätter)
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, ≥150 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from Bacillus amyloliquefaciens, liquid, ≥250 units/g
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, aqueous solution, ≥800 FAU/g
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~30 U/mg
Supelco
α-Amylase from Bacillus licheniformis, suitable for determination of starch (Kit STA-20)
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, Type XII-A, saline solution, ≥500 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, lyophilized powder, 500-1,500 units/mg protein, 93-100% (SDS-PAGE)
Sigma-Aldrich
α-Amylase from human saliva, Type XIII-A, lyophilized powder, 300-1,500 units/mg protein
Sigma-Aldrich
α-Amylase from human saliva, Type IX-A, lyophilized powder, 1,000-3,000 units/mg protein